Learn More
Tocris Bioscience™ Mibefradil dihydrochloride
CaV3.x blocker
Brand:  Tocris Bioscience™ 2198/50
922.52 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18093982
  • € 1175.00 / 50mg
Explore more special offers
Description
Description
Mibefradil dihydrochloride is a Ca2+ channel blocker with moderate selectivity for T-type Ca2+ channels displaying IC50 values of 2.7 μM and 18.6 μM for T-type and L-type channels respectively. Antihypertensive agent.
Specifications
Specifications
>97%
Mibefradil dihydrochloride
Mibefradil dihydrochloride
50 mg
568.56