Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
For use with V20 Karl Fischer Autotitrators.
Brand: Mettler Toledo™ 00023961
This item is not returnable.
View return policy
Description
- Accessory for DV1000 titration burettes
- For titrant protection from moisture or CO2
- With cover
Specifications
Drying Cup | |
Water content determination | |
453.5 g |