Learn More
Mettler Toledo™ Draining Adapter for V20 and V30 Series Karl Fischer™ Titrators
For use with V20 and V30 Karl Fischer titrators.
Brand:  Mettler Toledo™ 51105594
Product Code. 12668189
  • € 42.05 / Each
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Description
  • For use with V20 and V30 Karl Fischer titrators
  • For automatic draining of cell
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Draining Adapter
Automatic draining of cell
V20 and V30 series Karl Fischer titrator
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ Draining Adapter for V20 and V30 Series Karl Fischer™ Titrators > Adapter for automatic draining of cell