Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
Product Code. 15947301
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Unit Size:
Each
Product Code. 15947301
Brand: Metro™ TTM21S
Only null left
Add to basket
Add to basket
Includes
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
High-density, easy access storage.
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Product Type Mobile Unit Kit
Finish None
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Weight (English) 33 lb.
Weight (Metric) 14.8 kg
Width (English) Shelf 21 in.
Width (English) 23.25 in.
Width (Metric) Shelf 53 cm
Width (Metric) 59.05 cm
Material Stainless Steel