Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Unit Size:
Each
Description
Includes
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Mobile units move along anchored track allowing easy access to all shelves
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Specifications
| Product Type | Mobile Unit Kit |
| Finish | Chrome |
| For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system |
| Includes | Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves |
| Weight (English) | 33 lb. |
| Weight (Metric) | 14.8 kg |
| Width (English) Shelf | 21 in. |
| Width (English) | 23.25 in. |
| Width (Metric) Shelf | 53 cm |
| Width (Metric) | 59.05 cm |
| Show More |