Learn More
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits
Product Code. 15917291
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Unit Size:
Each
Product Code. 15917291
Brand: Metro™ TTE18K3
Only null left
Add to basket
Add to basket
Connects stationary end units to overhead track sets.
  • Kit includes 86 in. (218.5cm) posts and hardware necessary for connecting stationary end units to track
  • Super Erecta Shelves open-wire design minimizes dust accumulation, allows a free circulation of air and greater visibility of stored items.
MetroSeal 3 Finish is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
One kit is requird per single system. Shelves sold separately.
Specifications
Product Type Stationary End Unit Kit
Finish Metroseal 3 with Microban™
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes 86 in. Posts and connecting hardware
Weight (English) 11 lb.
Weight (Metric) 4.9 kg
Width (English) Shelf 18 in.
Width (English) 18 in.
Width (Metric) Shelf 45.7 cm
Width (Metric) 45.7 cm
Material Steel