Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
Brand: Metro™ 2460NFS
Description
- All-stainless construction
- Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Solid Shelf | |
Stainless Steel | |
60 x 24 in. (152.4 x 61 cm) | |
60 in. | |
24 in. |
Silver | |
Autoclavable | |
Metro Super Erecta Shelving System | |
152.4 cm | |
61 cm |