Learn More
Tocris Bioscience™ Methylmalonate
Succinate dehydrogenase inhibitor
Brand:  Tocris Bioscience™ 4979/50
69.98 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18784857
Description
Description
Methylmalonate is an intracellularly produced malonate, a reversible succinate dehydrogenase inhibitor used to generate Huntington's disease models. Induces apoptotic cell death in striatal neurons.
Specifications
Specifications
>98%
Methylmalonate
Methylmalonate
50 mg
118.09