Learn More
Methylgermanium trichloride, 97%
Product Code. p-7051527
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
Product Code. 11394168
Brand: Thermo Scientific Alfa Aesar 041449.02
Request Bulk or Custom Format
CAS: 993-10-2 | CH9Cl3Ge | 200.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylgermanium trichloride is used as a precursor for the synthesis of five-coordinated spirocyclic anionic methylgermanates by reacting with catechol. It is also involved in the photo-chemical adsorption of organo-germanium on a hydrogen terminated atomically flat silicon surface using xenon excimer light.
Solubility
Miscible with ether.
Notes
Sensitive to moisture and reacts violently with water. Also hydrolysis takes place when contact with water. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 993-10-2
Molecular Formula CH9Cl3Ge
Molecular Weight (g/mol) 200.06
MDL Number MFCD00013585
InChI Key NNMJXXWNUALEKD-UHFFFAOYSA-N
Synonym methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
PubChem CID 70436
IUPAC Name trichloro(methyl)germane
SMILES Cl.Cl.Cl.C[GeH3]
Specifications
Color Colorless to Yellow
Density 1.706 g/mL
Boiling Point 111°C
Flash Point 10°C (50°F)
Odor Chlorine-like
Linear Formula CH3GeCl3
Refractive Index 1.4685
Quantity 0.5 g
UN Number UN2924
Sensitivity Moisture sensitive
Formula Weight 193.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylgermanium trichloride