Learn More
Methylene Chloride, Acidified EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12950464 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 12950464
Brand: Fisher Chemical RP 128 100
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Packaging Bottle
Analyte or Component Names Methylene Chloride, Acidified.
Purity Grade Notes EU Ph Ref: 1055901
Concentration 0.1% in alcohol: water - 20:80
Element Methylene Chloride
Quantity 100 mL
Grade Eu. Ph.
compliance-icons
Product Identifier
  • Methylene Chloride,Acidified. European Pharmacopoeia Ref: 1055901 (RP 128)
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .