missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, ACS reagent

Product Code. p-3756597 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10311501

Brand: Thermo Scientific Chemicals 423970100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 178.0°C to 182.0°C
Clarity Passes Test
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
Beilstein 16, 329
Merck Index 15, 6192
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol, acetic acid
Infrared Spectrum Authentic
Formula Weight 269.3
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 4.2 (pink) to pH 6.2 (yellow)
Color Purple to Red
Quantity 10 g
Physical Form Powder
Chemical Name or Material Methyl Red, Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.