Learn More
MedChemExpress Methyl protodioscin
MedChemExpress Methyl protodioscin(NSC-698790) is a furostanol bisglycoside with antitumor properties; shows to reduce proliferation, cause cell cycle arrest.
Brand:  MedChemExpress HY-N0863
Encompass_Preferred
Product Code. 16491548
  • € 321.30 / 5mg
Explore more special offers
Specifications
4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Room temperature
54522-52-0
98.0%
NSC-698790∣ Smilax saponin B
5 mg
1063.23 g/mol
Cancer-programmed cell death
Natural Products
DMSO : 140 mg/mL (131.67 mM∣Need ultrasonic) ∣H2O : 25 mg/mL (23.51 mM∣Need ultrasonic)
Apoptosis
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers