Learn More
Lauda™ Insulated Hose
Material: Metal
Brand: Lauda™ LZM 044
This item is not returnable.
View return policy
Description
- For connections with thread M16x1
Specifications
10°C to 400°C | |
Metal | |
18 mm |
Hose | |
20 cm | |
10 mm |