missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human MET (cMET), His Tag Recombinant Protein
Human MET (cMET), His Tag Recombinant Protein
€ 596.00 - € 7440.00
Specifications
Accession Number | NP_000236.2 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 53.7 kDa |
Gene Alias | MET (cMET) |
KinaseFamily | MET (HGFR) Kinase Family |
Product Code | Brand | Description | Price | Quantity | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity | Availability | ||||
11549326
|
Thermo Scientific™
PV3143 |
10μg | 10μg |
€ 596.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073172
|
Thermo Scientific™
PV3588 |
1 mg | 1 mg |
€ 7440.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MET (cMET) is a receptor-like tyrosine kinase whose disregulation has been linked to many types of human malignancies.Specifications
NP_000236.2 | |
53.7 kDa | |
MET (HGFR) Kinase Family | |
Recombinant | |
Storage Buffer: 20 mM Tris, pH 7.5, 100 mM NaCl, 0.5 mM EDTA, 0.05% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
MET (cMET) | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus |