missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury ICP Standard, 1000 mg/L Hg in HNO3 10%, Honeywell Fluka™
Brand: Honeywell Fluka E1026-100ML
This item is not returnable.
View return policy
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Mercury ICP Standard | |
100 mL | |
Standards for ICP and ICP-MS | |
<2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction