missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) Chloride, ≥99.5%, ACS Reagent, Honeywell Fluka™

Product Code. 15602300
Click to view available options
Quantity:
5 g
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.084
MDL Number MFCD00011043
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]

Specifications

Melting Point 400°C
Color White
Quantity 5 g
Linear Formula Hg2Cl2
UN Number UN2025
Formula Weight 472.09g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 7.15g/cm3
Chemical Name or Material Mercury(I) Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'