missing translation for 'onlineSavingsMsg'
Learn More

Haleco™ Mercury Collection Kit

Contains all of the basic elements needed for the collection of a spill or leak of mercury.

Brand:  Haleco™ CKT0000MZ


Product Code. 11965295

  • € 154.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 2 absorbent sponges
  • 1 collection box for sponges with screw cap
  • 1 bottle of 500g of absorbent powder
Specifications

Specifications

Mercury Cleaning Kit
Cleaning mercury spills and leaks
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • mercury recovery kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : zinc

SDS
Documents

Documents

Certificates
Special Offers

Special Offers