missing translation for 'onlineSavingsMsg'
Learn More

Mepyramine maleate, Tocris Bioscience™

Product Code. 18722634
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective H1 inverse agonist

Selective inverse agonist for the H1 receptor. Inhibits histamine induced inositol phosphate (InsP) production (log EC50 = -7.94) and intracellular calcium mobilization. Sequesters Gq/11 protein, reducing its availability for other receptors associated with the same signaling pathway.

Specifications

CAS 59-33-6
Molecular Formula C21H27N3O5
MDL Number MFCD00069333
Quantity 100 mg
Synonym pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
InChI Key JXYWFNAQESKDNC-BTJKTKAUSA-N
SMILES OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1
IUPAC Name (2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Molecular Weight (g/mol) 401.46
PubChem CID 5284451
Formula Weight 401.46
Percent Purity >98%
Chemical Name or Material Mepyramine maleate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'