Learn More
B Braun™ Surface Spray Disinfectant
Use B Braun™ Surface Spray Disinfectant for rapid surface disinfection with Rapid Meliseptrol. The broad-spectrum disinfectant has a 5-minute reaction time.
Brand:  B Braun™ 18565
 View more versions of this product
Product Code. 10381722
  • € 26.00 / 1 L
Explore more special offers
Description
Description
  • Rapid Meliseptrol distribution
  • Free of CFC's
  • For small surfaces and equipment
  • Colorless
Order Info
VolumeMetric
Specifications
Specifications
Disinfectant
Liquid
1 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
B Braun™ Surface Spray Disinfectant > Capacity: 1L