Learn More
Thermo Scientific Chemicals Melatonine, 99%
Melatonine, 99%, CAS # 73-31-4, is a hormone usually produced in the pineal gland and released into the blood.
€ 47.64 - € 350.75
Chemical Identifiers
CAS | 73-31-4 |
---|---|
Molecular Formula | C13H16N2O2 |
Molecular Weight (g/mol) | 232.28 |
MDL Number | MFCD00005655 |
InChI Key | DRLFMBDRBRZALE-UHFFFAOYSA-N |
Synonym | melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovine |
PubChem CID | 896 |
ChEBI | CHEBI:16796 |
IUPAC Name | N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide |
SMILES | CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10255030
|
Thermo Scientific Chemicals
125362500 |
250 mg | Glass bottle |
€ 47.64
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316960
|
Thermo Scientific Chemicals
125360010 |
1 g | Glass bottle |
€ 100.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267063
|
Thermo Scientific Chemicals
125360050 |
5 g | Glass bottle |
€ 350.75
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Melatonine is a hormone produced by the pineal gland that shows several biological activities, including the somnolence, sleep-wake cycle, circadian rhythm, stress response, aging, and immunity activities
• It shows antioxidant, autocoid, paracoid, hormonal, anti- inflammatory and anti-apoptotic activities
Applications
• Melatonin shows neurohormonal activity and can upregulate antioxidant enzymes and inhibit rat NOS1
• Melatonin has been reported to be an activator of MEL-1A-R and MEL-1B-R
• The production and secretion of melatonine are dependent on beta-adrenergic receptor function
• During melatonine synthesis, the essential amino acid L-tryptophan is used as a precursor
• This compound inhibits apoptosis in immune cells and neurons, and enhances apoptotic cell death of cancer cells
• It can inhibit proliferation and metastasis of breast cancer cells by modulating Rho-associated kinase protein-1 expression
Chemical Identifiers
73-31-4 | |
232.28 | |
DRLFMBDRBRZALE-UHFFFAOYSA-N | |
896 | |
N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide |
C13H16N2O2 | |
MFCD00005655 | |
melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovine | |
CHEBI:16796 | |
CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC |
Specifications
73-31-4 | |
Cream to Yellow or White | |
Authentic | |
C13H16N2O2 | |
22, V,12, 42 | |
melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovine | |
DRLFMBDRBRZALE-UHFFFAOYSA-N | |
N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide | |
896 | |
232.28 | |
Powder |
116.0°C to 120.0°C | |
0.5% max. (1 g, 105°C) | |
98.5% min. (HPLC) | |
MFCD00005655 | |
15, 5884 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene,toluene,hexane,acet,onitrile | |
CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC | |
232.28 | |
CHEBI:16796 | |
99% | |
Melatonine |
Safety and Handling
missing translation for 'einecsNumber' : 200-797-7
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.