Learn More
GA International Medium Round Cryogenic Writable Labels
Writable cryogenic color dots provided in dispenser box rolls for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen Dewars.
€ 34.17
Specifications
Adhesive Type | Permanent |
---|---|
Finish | Matte |
Format | Roll |
Material | Thermoplastic Film |
Product Type | Labels |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17809187
|
GA International
JTRA-11BE |
Beige |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17828977
|
GA International
JTRA-11BR |
Brown |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17881308
|
GA International
JTRA-11GA |
Green |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17888977
|
GA International
JTRA-11GS |
Seafoam |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17829187
|
GA International
JTRA-11LA |
Lavender |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870548
|
GA International
JTRA-11LE |
Lemon |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810068
|
GA International
JTRA-11MT |
Mint |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17899367
|
GA International
JTRA-11OR |
Orange |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870268
|
GA International
JTRA-11RE |
Red |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17819577
|
GA International
JTRA-11SI |
Silver |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17820838
|
GA International
JTRA-11VI |
Violet |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17871078
|
GA International
JTRA-11YE |
Yellow |
€ 34.17
Pack of 1100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Strong adhesive ensures labels won't wrinkle or peel during long-term storage in -80°C freezers, liquid nitrogen and dry ice
- Available in a wide range of colors, for color coding and easy identification
- Small round labels are ideal for Eppendorf tube caps and vials
- Available in a variety of colors
- Writable with permanent cryo markers
Specifications
Permanent | |
Roll | |
Labels | |
2 Labels | |
Round | |
-196°C to +110°C | |
1 in. |
Matte | |
Thermoplastic Film | |
Non-sterile | |
1100/Pk. | |
-321°F to +230°F | |
11 mm | |
25.4 mm |