Learn More
Liebherr™ Spark-Free interior, Lab Fridge
Product Code. 11986249 Shop All Liebherr Products
Click to view available options
Capacity (Metric):
180 L
180 L (gross)
333 L
554 L
Style:
Table-height
Tall
Unit Size:
Each
Product Code. 11986249
Brand: Liebherr™ LKEXV1800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For secure, accurate and long-lasting, spark-free interior, fridge storage of non temperature-sensitive products, the Liebherr™ Spark-Free Interior, Lab Fridge is designed for general purpose applications with Liebherr build-quality you can rely on.
  • Spark-Free Lab Fridge for storage of solvents and reagents, reach-in, 1-section, width 600mm.
  • Temperature range +1 / +15°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
  • Climate class 4, refrigerant R 600a, connection rating 1,0A, 230v/50/1-ph,energy consumption 328 kWh/a
  • This model is intended for the spark-free storage of chemicals where a high level of temperature stability is not required
  • ATEX-certified spark-free interior
  • Analogue controls with digital temperature display in the door.
  • Door lock, reversible hinges
  • 40 kg shelf load
Specifications
Type Laboratory Refrigerator
Capacity (Metric) 180 L
Temperature Range 1°C to 15°C
Temperature Resolution 1.0°C
Temperature Stability Gradient 10.2°C
Refrigerant R 600a
Door Style Solid
No. of Doors 1
Hinge Side Right, reversible
No. of Drawers 0
Shelves 3 Glass
Capacity (Metric) Shelf 40 kg
Cabinet Material Steel
Exterior Finish White-coated Steel
Insulation 40 mm HDPE
Interior Polystyrol
Certifications/Compliance ATEX Certified
Style Table-height
Display Digital, External
Color White
Control Type Analog
Defrost Automatic
Compressor Forced-air
Depth (Metric) Exterior 60 cm
Depth (Metric) Interior 44.1 cm
Height (Metric) Exterior 86 cm
Height (Metric) Interior 70.2 cm
Width (Metric) Exterior 60 cm
Width (Metric) Interior 51.3 cm
Amperage 1.0 A
Voltage 220/240 V
Electrical Requirements 220 to 240 V 50 Hz 1.0 A
Frequency 50 Hz
Energy Usage 328 kWh/a
Shipping Weight (Metric) 41 kg
Weight (Metric) 38 kg