Learn More
Medical Packaging Cytology Brush
Use to collect cervical cells for Pap smear and/or STD testing
Brand:  Medical Packaging 22-281-660
Encompass_Preferred
 View more versions of this product
Product Code. 13436626
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Soft nylon bristles increases collection of endocervical cells and component material
Specifications
Specifications
Cytology Brush
Sterile
15.2 cm
100 Pk.
Nylon
6 in.
Use to collect cervical cells for Pap smear and STD testing
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Medical Packaging Cytology Brush