missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MBPTrap™ HP Prepacked Columns
Shop All Cytiva Products
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
5 x 5mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
1mL
Each
Pack of 5
Description
- Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
- Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
- Maximum pressure: 3 bar
- Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
- 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
| Buffer | Aqueous Buffer |
| Product Type | Dextrin Sepharose HP prepacked column |
| Bed Volume | 1mL |
| Diameter (English) Inner | 0.27 in. |
| Diameter (Metric) Inner | 7mm |
| Quantity | 1 x 1mL |
| Height (English) | 0.98 in. |
| Height (Metric) | 25mm |
| Flow Rate | <4 mL/min. |
| Material | Polypropylene |
| Show More |
Safety and Handling
Product Identifier
- MBPTrap HP, 1 ml, 1 x 1 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction