Learn More
MBL Volumetrics™ Glass Graduated Cylinder
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.1 mL
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 2
Description
Manufactured from chemically resistant borosilicate glass. MBL Volumetrics™ Glass Graduated Cylinders are a reliable choice for measuring liquids and solutions in the laboratory.
Features
- Easy to read graduations in blue enamel
- Sturdy hexagonal foot
- Complies with ISO 4788
Specifications
Specifications
| Base Type | Hexagonal |
| Scale Color | Blue |
| Shape | Round |
| Type | Graduated Cylinder |
| Tolerance | ±20 mL |
| Material | Borosilicate Glass |
| Certifications/Compliance | DIN 12680, ISO 4788, BS 604 |
| Class | B |
| Capacity (Metric) | 2000 mL |
| For Use With (Application) | Measuring Liquids and Solutions in the Laboratory |
| Show More |