Learn More
Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Dual Stacked Universal Platform w/o Clamps
Platform w/o Clamps
Stacking Adapter
Universal Platform w/o Clamps
For Use With (Equipment):
MaxQ 2000
MaxQ 2000, 2506, 2508, 4000, 6000
MaxQ 2000, 2506, 2508, 4450
MaxQ 2000, 6000
MaxQ 3000
MaxQ 3000, 5000
MaxQ 30100, MaxQ 30110 Platform
MaxQ 7000
Dimensions (L x W):
11 x 13 in.
13 x 11 in.
18 x 18 in.
18 x 30 in.
24 x 36 in.
Unit Size:
Each
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ 30100 platforms (11 × 13in.) and MaxQ 30110 platforms (18 × 18in.).
Specifications
Specifications
| Product Type | Stacking Adapter |
| Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
| Dimensions (L x W) | 13 x 11 in. |
| Length (English) Platform | 11 in. (MaxQ 30100 Platforms), 18 in. (MaxQ 30110 Platforms) |
| Length (Metric) Platform | 279.4 mm (MaxQ 30100 Platforms), 457.2 mm (MaxQ 30110 Platforms) |
| For Use With (Equipment) | MaxQ 30100, MaxQ 30110 Platform |