Learn More
Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Dual Stacked Universal Platform w/o Clamps
Platform w/o Clamps
Stacking Adapter
Universal Platform w/o Clamps
For Use With (Equipment):
MaxQ 2000
MaxQ 2000, 2506, 2508, 4000, 6000
MaxQ 2000, 2506, 2508, 4450
MaxQ 2000, 6000
MaxQ 3000
MaxQ 3000, 5000
MaxQ 30100, MaxQ 30110 Platform
MaxQ 7000
Dimensions (L x W):
11 x 13 in.
13 x 11 in.
18 x 18 in.
18 x 30 in.
24 x 36 in.
Unit Size:
Each
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Specifications
| Product Type | Universal Platform w/o Clamps |
| Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
| Dimensions (L x W) | 24 x 36 in. |
| Length (English) Platform | 36 in. |
| Length (Metric) Platform | 91.4 cm |
| Width (English) Platform | 24 in. |
| Width (Metric) Platform | 60.9 cm |
| For Use With (Equipment) | MaxQ 3000 |