Learn More
Masterflex™ Translucent C-Flex Braided Tubing
Click to view available options
Diameter (Metric) Outer:
12.7 mm
19.05 mm
Unit Size:
25ft
Each
Specifications
Specifications
| Certifications/Compliance | FDA compliant, and complies with USP Class VI standards |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 6.4 mm |
| Material | C-Flex |
| Diameter (Metric) Outer | 12.7 mm |
| Overall Length (Metric) | 7.62 m |
| For Use With (Application) | For use with cell cultures, vessel and tank transfer, pump applications and cold storage. |
| Min. Operating Temperature (Metric) | -45°C |
| Packaging Quantity | 1 Pack |
| Color | Translucent |
| Show More |