Learn More
RL Enterprises™ Mass Slotted
Brand:  RL Enterprises™ MAS-704-030N
Product Code. 12846425
  • € 14.54 / Pack of 5
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Mass Slotted
100 g
5 Pack
5 Pack
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RL Enterprises™ Mass Slotted > Mass: 100g