Learn More
Emma Safety Footwear Marco (Frontier 112) Safety Shoes
Made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
Brand:  Emma Safety Footwear MM11209044
 View more versions of this product
Product Code. 15785233
  • € 241.00 / 1 pair
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Thermoplastic Polyurethane Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver & Wellness leather
44
Laces
Marco (Frontier 112)
Leather
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear Marco (Frontier 112) Safety Shoes > Size: 44