missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.
Solubility
Soluble to 50mg/ml in water. Soluble in ethanol at 10mM
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.
Solubility
Soluble to 50mg/ml in water. Soluble in ethanol at 10mM
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Specifications
Specifications
Quantity | 5g |
Beilstein | 5463242 |
Merck Index | 14,5748 |
Solubility Information | Soluble to 50mg/ml in water; Soluble in ethanol at 10mM |
Formula Weight | 313.87 |
Percent Purity | 99% |
Chemical Name or Material | Maprotiline hydrochloride |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X03K004 | Certificate of Analysis | 16/10/2024 | J62321.06,J62321.03 |
W14C066 | Certificate of Analysis | 08/12/2021 | J62321.06,J62321.03 |
W08B042 | Certificate of Analysis | 08/12/2021 | |
U30F005 | Certificate of Analysis | 08/12/2021 | J62321.06,J62321.03 |
U12B035 | Certificate of Analysis | 07/12/2021 | J62321.06,J62321.03 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Maprotiline hydrochloride, 99%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction