Learn More
SI Analytics™ Manual Ubbelohde Viscometer
Product Code. 12048163 Shop All SI Analytics Products
Click to view available options
Approximate Constant:
0.001K
0.003K
0.01K
0.03K
0.05K
0.1K
0.3K
1K
3K
Size:
0.36mm Capillary Diameter
0.47mm Capillary Diameter
0.63mm Capillary Diameter
0.84mm Capillary Diameter
0.95mm Capillary Diameter
1.13mm Capillary Diameter
1.50mm Capillary Diameter
2.01mm Capillary Diameter
2.65mm Capillary Diameter
Unit Size:
Each
Product Code. 12048163
Brand: SI Analytics™ 285400061
Only null left
Add to basket
Add to basket
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Manual Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated with constant for manual measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Calibrated Yes
Description 290mm Long
Approximate Constant 0.1K
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Includes Ring Marks
For Use With (Application) For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 1.13mm Capillary Diameter
Volume (Metric) 15 to 25 mL