Learn More
Thermo Scientific™ Manual GC Syringes
Product Code. p-4525911 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
Needle Gauge:
23 G
23s G
25 G
26 G
Product Type:
GC Autosampler Syringe, Fixed Needle
GC Autosampler Syringe, Plunger-in-Needle, 50 mm
GC Autosampler Syringe, Plunger-in-Needle, 70 mm
GC Autosampler Syringe, Removable Needle
Quantity:
6/Pk.
Ea.
Unit Size:
Each
Pack of 6
Product Code. 10361195
Brand: Thermo Scientific™ 365B0511
Easily accomplish accurate, precise, and reproducible manual injections with Thermo Scientific™ GC Syringes.
Achieve accurate, precise, and reproducible manual injections easily using Thermo Scientific™ Manual GC Syringes. The robust, field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes. Easy-to-read markings ensure reproducibility.
  • Syringes available in a range of volumes
  • PTFE Luer-Lok™ syringes
  • PTFE Luer Slip syringes
TRUSTED_SUSTAINABILITY
Specifications
Description 1.0uL; 50mm L; Plunger-in-needle
Needle Gauge 23 G
Length (Metric) 50 mm
Product Type GC Autosampler Syringe, Plunger-in-Needle, 50 mm
Disposable Reusable
Quantity Ea.
Volume (Metric) 1 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) GC Autosampler
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.