Learn More
Macherey-Nagel™ Manual Crimper
Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
11 mm Aluminum Crimp Caps
13 mm Aluminum Crimp Caps
20 mm Aluminum Crimp Caps
8 mm Aluminum Caps
Unit Size:
Each
Specifications
Specifications
| Type | Manual Crimper |
| Quantity | 1 Each |
| For Use With (Equipment) | 20 mm Aluminum Crimp Caps |