Learn More
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand:  Value Plastics 50120-95
 View more versions of this product
Product Code. 30053779
  • € 35.71 / Pack of 25
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
1/16 in.
Polypropylene
Male Luer x Hose Barb
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb > 1/16 in.,Polypropylene
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.