Learn More
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand:  Value Plastics 50120-23
 View more versions of this product
Product Code. 30053606
  • € 24.14 / Pack of 25
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
Non-DEHP;Non-Phthalate;RoHS
3/32 in.
Nylon
Male Luer x Hose Barb
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb > 3/32 in.,Nylon
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.