Learn More
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand: Value Plastics 50120-27
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtimeSpecifications
Non-DEHP;Non-Phthalate;RoHS;FDA | |
1/8 in. | |
Polycarbonate | |
Male Luer x Hose Barb |
Product Content Correction
Product Title
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb > 1/8 in.,Polycarbonate
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.