Learn More
Buerkle™ Magnetic Stirring Bar
Product Code. 10581063 Shop All Bürkle Products
Click to view available options
Length (Metric):
20 mm
25 mm
30 mm
40 mm
50 mm
80 mm
Diameter (Metric):
6 mm
7 mm
8 mm
10 mm
Unit Size:
Each
Product Code. 10581063
Brand: Buerkle™ 82712006
Only null left
Add to basket
Add to basket
Molded with an outer layer of chemically inert PTFE for durability and non-reactivity. Buerkle™ Magnetic Stirring Bars feature a cylindrical shape and smooth surface for excellent mixing capability.
  • Opaque PTFE construction
  • High performance permanent magnets 
  • Long lifespan
  • Exceptional temperature resistance
  • Wide range of sizes
  • Stirring bars with diameters of 6 and 8mm can be used with Buerkle™ Fish-clip™ for extra gentle stirring
TRUSTED_SUSTAINABILITY
Specifications
Color White
Diameter (Metric) 6 mm
Length (Metric) 20 mm
Shape Cylinder