Learn More
Velp Scientifica™ Magnetic Stir Bars
Product Code. 15895221 Shop All Velp Products
Click to view available options
Length (Metric):
5 mm
6 mm
8 mm
10 mm
13 mm
30 mm
Diameter (Metric):
8 mm
10 mm
20 mm
Unit Size:
Each
Product Code. 15895221
Brand: Velp Scientifica™ A00000356
Only null left
Add to basket
Add to basket
Velp Scientifica™ Magnetic Stir Bars for use with Velp Scientifica™ Hotplate Stirrers.
  • For use with Velp Hotplate Stirrers
  • Available in various sizes and shapes
Specifications
For Use With (Equipment) Hotplate Stirrers
Diameter (Metric) 8 mm
Length (Metric) 30 mm
Shape Cylinder