Specifications
ISO 7010 | |
Plastic | |
Permanent Acrylic | |
297 mm | |
-18°C to 70°C | |
Flammable Gases 2 | |
Black on Red |
ADR 2.1A-297X297-B859 | |
1 Ea. | |
Magnetic Plastic | |
297 mm | |
Rhomboid | |
Dangerous Goods Transport Label |