missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
100 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 4 x 5 kg |
Linear Formula | MgSO4 · 7H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 246.47 |
Percent Purity | 99.5 to 100.5% (calc. to the dried substance) |
Vapor Pressure | 0.1mmHg (20°C) |
Density | Vapor Density: <0.01 (vs air) |
Chemical Name or Material | Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0459716 | Certificate of Analysis | 01/02/2024 | 131420250,131420010,13142-1KG,131421000,13142-4X5KG,13142-5KG,13142-25KG |
A0391837 | Certificate of Analysis | 15/12/2022 | 131420250,131420010,13142-1KG,131421000,13142-4X5KG,13142-5KG,13142-25KG |
A0398507 | Certificate of Analysis | 12/12/2022 | 131420250,131420010,13142-1KG,131421000,13142-4X5KG,13142-5KG,13142-25KG |
A0429770 | Certificate of Analysis | 09/11/2022 | 131420250,131420010,13142-1KG,131421000,13142-4X5KG,13142-5KG,13142-25KG |
A0446422 | Certificate of Analysis | 07/10/2022 | 131420250,131420010,13142-1KG,131421000,13142-4X5KG,13142-5KG,13142-25KG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction