missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate Hexahydrate, Honeywell Fluka™

Product Code. 15682360
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate

Specifications

Quantity 100 g
Linear Formula Mg(NO3)2 · 6H2O
UN Number NONH for all modes of transport
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
Formula Weight 256.41g/mol
Percent Purity 99%
Grade ACS Reagent
Density Vapor Desnity: 6 (vs air)
Chemical Name or Material Magnesium Nitrate Hexahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'