missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydroxide, 95-100.5%, Thermo Scientific Chemicals

Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes

€ 32.16 - € 180.90

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11305929
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012367.22
100 g
€ 32.16
100g
Please to purchase this item. Need a web account? Register with us today!
11331759
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012367.36
500 g
€ 47.13
500g
Please to purchase this item. Need a web account? Register with us today!
11315929
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012367.A4
2.5 kg
€ 180.90
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as an additive in certain refractories. It is applied as a non-hazardous alkali in neutralizing acidic wastewater. It is also used in making artificial reefs. It widely used in milk of magnesia antacids and laxatives. Also works as an antiperspirant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1309-42-8
58.32
VTHJTEIRLNZDEV-UHFFFAOYSA-L
73981
[OH-].[OH-].[Mg++]
H2MgO2
MFCD00011104
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
CHEBI:6637

Specifications

360°dec
White
95-100.5%
MFCD00011104
Soluble in dilute acids. Slightly soluble in water.
[OH-].[OH-].[Mg++]
58.32
CHEBI:6637
Odorless
White powder
Magnesium hydroxide
1309-42-8
Powder
H2MgO2
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
VTHJTEIRLNZDEV-UHFFFAOYSA-L
magnesium;dihydroxide
73981
58.33
Air Sensitive
2.36
Videos
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 215-170-3

missing translation for 'rtecsNumber' : OM3570000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only