Learn More
Thermo Scientific Chemicals Magnesium chloride hexahydrate, 99%, for biochemistry Product Code.: 13226319
Thermo Scientific Chemicals Magnesium chloride hexahydrate, 99%, for biochemistry
Product Code. 13226319
2.5 kg
Click to view available options
Quantity:
500 g
2.5 kg
Product Code. 13226319
Brand: Thermo Scientific Chemicals 447150025
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
Specifications
Melting Point 117.0°C
Quantity 2.5 kg
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula MgCl2·6H2O
Merck Index 15,5726
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 203.31
Percent Purity 99%
Grade Biochemical
Chemical Name or Material Magnesium chloride hexahydrate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460941 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number447150025,447155000
Lot NumberA0456583 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number447150025,447155000
Lot NumberA0456582 Certificate TypeCertificate of Analysis Date27/09/2023 Catalog Number447150025,447155000
Lot NumberA0410579 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number447150025,447155000
Lot NumberA0453984 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number447150025,447155000
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Magnesium chloride hexahydrate, 99%, for biochemistry >