Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit
Product Code. 16681236
Click to view available options
Quantity:
Up to 100 Preps
Up to 1000 Preps
Unit Size:
1 set
1000 reactions
Product Code. 16681236
Brand: Applied Biosystems™ A48310
The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM).
The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM). The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing.
Features of the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit include:
• Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
• Fast procedure allows for 96 samples to be processed in under 30 minutes
• Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of biofluids or transfer media
• No need for carrier RNA
• Elution volumes ranging from 50 to 100 μL
Maximum sample input in a 96-deep well or 24-deep well plate format
The optimized reagents included in the MagMAX Viral/Pathogen kit allow you to maximize the amount of sample input, thereby increasing the amount of RNA and/or DNA recovered. Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate. For larger sample volumes, protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate.
Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.
The Dynabeads MyOne Silane contained in the MagMAX Viral/Pathogen Total Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with a highly specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery viral RNA and DNA present at low concentrations in body fluids. Once captured, these nucleic acids can then be eluted in 50 μL of elution buffer for use in downstream applications.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage • 550 mL Binding Solution, store at room temperature
• 1000 mL Wash Solution, store at room temperature
• 100 mL Elution Solution, store at room temperature
• 10 mL Proteinase K, store at room temperature
• 20 mL DNA/RNA Binding Beads, store at room temperature
Format 96 Deep-well Plate, 24 Deep-well Plate
Purification Time 30 min.
Product Line MagMAX
Quantity Up to 1000 Preps
Shipping Condition Room Temperature