missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady Magicard™ Cleaning Kit for the Prima 8
Click to view available options
Unit Size:
Each
Description
- Ensure high quality printing on every card.
- Good for 10 cleaning cycles.
- Kit holds cleaning cards and cleaning pen.
Specifications
Specifications
| Product Type | Cleaning Kit |
| For Use With (Equipment) | Prima 8 |
| Description | Cleaning Kit (10 pads + cards) for Prima 8 Printer |
| Format | Cleaning Kit |
| Includes | 10 Cleaning Pads and Cards |
| For Use With (Application) | Healthcare and Hospital labelling;General identification;Facility identification;Security identification |
| Quantity | 1 Ea. |
| Alternate Part Number | E9887 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction