missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix C, Bulk
Lysing Matrix C contains 1 mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast, fungi, molds, spores and filamentous algae.
Brand: MP Biomedicals™ 116540433
This item is not returnable.
View return policy
Description
- Product Overview: 1 mm silica spheres. Found in the FastRNA Pro Red Kit and FastProtein Red Matrix. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
- Applications: Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
Specifications
2°C to 8°C | |
Yeast, Fungi, Algae | |
Lysing Matrix C | |
1 kg |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.