Learn More
Abnova™ LY6E 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00004061-T02.100uL
Product Code. 16147481
  • € 215.00 / 100µL
Explore more special offers
Specifications
293T
LY6E 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 13.5kDa
Transient overexpression cell lysate was tested with anti-LY6E antibody (Mfr. No. H00004061-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_002346.1
pCMV-LY6E full-length
100 μL
Human
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abnova™ LY6E 293T Cell Transient Overexpression Lysate (Denatured) > 100μL