Learn More
SI Analytics™ Low Maintenance pH Combination Electrode with Screw Plug Head
Shop All SI Analytics Products
Click to view available options
Length (Metric):
120 mm
225 mm
320 mm
425 mm
Junction:
Ceramic Diaphragm
KPG Annular Gap Diaphragm
Unit Size:
Each
Description
- Proven high-end electrodes for demanding measurement tasks
- Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Specifications
Specifications
| Connector Types | Screw Plug Head |
| Junction | KPG Annular Gap Diaphragm |
| Material | H-Glass |
| pH Range | 2 to 13 |
| Type | Low Maintenance pH Combination Electrode |
| Internal Reference | Silamid™ |
| Membrane | Dome |
| Model | H 8281 Hd |
| Temperature (Metric) Range | 0°C to 100°C |
| Length (Metric) | 225 mm |
| Show More |