Learn More
Brand™ Low-Form Polypropylene Beakers with Blue Printed Scale
Product Code. 15367864 Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
400 mL
500 mL
600 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Pack of 12
Pack of 4
Pack of 6
Product Code. 15367864
Brand: Brand™ 87622
Only null left
Add to basket
Add to basket
High clarity
  • With beaded rim and spout
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2000 mL
Material Polypropylene
Increments 20mL
Graduated Yes